Assay Details
| Gene Symbol: Nck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF043259.1 | - | 1 - 2 | 152 | 154 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF043259.1 transcripts.
Set Membership: |
|

Nck1
Genomic Map