Assay Details
| Gene Symbol: Nfatc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136073.2 | NP_001129545.1 | 6 - 7 | 2031 | 67 | |
| NM_001291168.1 | NP_001278097.1 | 6 - 7 | 1916 | 67 | ||
| NM_001291169.1 | NP_001278098.1 | 6 - 7 | 1916 | 67 | ||
| NM_001291170.1 | NP_001278099.1 | 6 - 7 | 1396 | 67 | ||
| NM_001291171.1 | NP_001278100.1 | 5 - 6 | 880 | 67 | ||
| NM_001291173.1 | NP_001278102.1 | 5 - 6 | 880 | 67 | ||
| NM_001291174.1 | NP_001278103.1 | 6 - 7 | 1396 | 67 | ||
| NM_001291175.1 | NP_001278104.1 | 6 - 7 | 1511 | 67 | ||
| NM_001291176.1 | NP_001278105.1 | 5 - 6 | 995 | 67 | ||
| NM_001291177.1 | NP_001278106.1 | 6 - 7 | 1511 | 67 | ||
| NM_001291178.1 | NP_001278107.1 | 5 - 6 | 995 | 67 | ||
| NM_010899.3 | NP_035029.2 | 6 - 7 | 2031 | 67 | ||
| GenBank mRNA | AK161174.1 | - | 4 - 5 | 606 | 67 | |
| EU887581.1 | - | 6 - 7 | 1797 | 67 | ||
| EU887582.1 | - | 6 - 7 | 1857 | 67 | ||
| EU887583.1 | - | 6 - 7 | 1394 | 67 | ||
| EU887584.1 | - | 6 - 7 | 1494 | 67 | ||
| EU887585.1 | - | 5 - 6 | 878 | 67 | ||
| EU887586.1 | - | 5 - 6 | 978 | 67 | ||
| EU887587.1 | - | 6 - 7 | 1797 | 67 | ||
| EU887588.1 | - | 6 - 7 | 1857 | 67 | ||
| EU887589.1 | - | 6 - 7 | 1394 | 67 | ||
| EU887590.1 | - | 6 - 7 | 1494 | 67 | ||
| EU887591.1 | - | 5 - 6 | 878 | 67 | ||
| EU887592.1 | - | 5 - 6 | 978 | 67 | ||
| U36575.1 | - | 6 - 7 | 1927 | 67 | ||
| U36576.1 | - | 6 - 7 | 1927 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc2
Genomic Map