| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800019760.mm.1 |
NM_001081982.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019760.mm.1 |
NM_010906.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019760.mm.1 |
TC0800002589.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019762.mm.1 |
NM_001081982.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019762.mm.1 |
NM_010906.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019762.mm.1 |
TC0800002589.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019761.mm.1 |
NM_001081982.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019761.mm.1 |
NM_010906.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019761.mm.1 |
TC0800002589.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019759.mm.1 |
NM_001081982.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019759.mm.1 |
NM_010906.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019759.mm.1 |
TC0800002589.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800019755.mm.1 |
NM_001081981.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019755.mm.1 |
NM_001081982.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019755.mm.1 |
NM_010906.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800019755.mm.1 |
TC0800002589.mm.1 |
MTA TC |