Assay Details
| Gene Symbol: Nfix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081981.2 | NP_001075450.1 | 5 - 6 | 1808 | 72 | |
| NM_001081982.2 | NP_001075451.1 | 5 - 6 | 1133 | 72 | ||
| NM_010906.3 | NP_035036.1 | 5 - 6 | 1133 | 72 | ||
| GenBank mRNA | BC003766.1 | - | 5 - 6 | 855 | 3491917 | 72 |
| BC090626.1 | - | 5 - 6 | 835 | 72 | ||
| S81451.1 | - | 4 - 5 | 819 | 72 | ||
| U57636.1 | - | 4 - 5 | 810 | 72 | ||
| Y07688.1 | - | 5 - 6 | 1135 | 72 | ||
| Y07689.1 | - | 5 - 6 | 937 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfix
Genomic Map