Assay Details
| Gene Symbol: Nfix | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297601.1 | NP_001284530.1 | 6 - 7 | 1942 | 64 | |
| NM_010906.3 | NP_035036.1 | 6 - 7 | 1270 | 64 | ||
| GenBank mRNA | AK158568.1 | - | 6 - 7 | 1945 | 64 | |
| U57636.1 | - | 5 - 6 | 947 | 64 | ||
| Y07689.1 | - | 6 - 7 | 1074 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfix
Genomic Map