Assay Details
| Gene Symbol: Nfya | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010913.2 | NP_035043.1 | 2 - 3 | 341 | 59 | |
| GenBank mRNA | AK143723.1 | - | 2 - 3 | 303 | 59 | |
| BC029695.1 | - | 2 - 3 | 229 | 5374597 | 59 | |
| M86214.1 | - | 1 - 2 | 78 | 59 | ||
| X55315.1 | - | 2 - 3 | 251 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfya
Genomic Map