Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800023434.mm.1 |
NM_010926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023434.mm.1 |
NM_010926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023434.mm.1 |
TC0800003249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023440.mm.1 |
NM_010926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023440.mm.1 |
NM_010926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023440.mm.1 |
TC0800003249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023437.mm.1 |
NM_010926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023437.mm.1 |
NM_010926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023437.mm.1 |
TC0800003249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023443.mm.1 |
NM_010926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023443.mm.1 |
NM_010926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023443.mm.1 |
TC0800003249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023438.mm.1 |
NM_010926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023438.mm.1 |
NM_010926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023438.mm.1 |
TC0800003249.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800023435.mm.1 |
NM_010926.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023435.mm.1 |
NM_010926.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800023435.mm.1 |
TC0800003249.mm.1 |
MTA TC |