Assay Details
| Gene Symbol: Nsdhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010941.3 | NP_035071.3 | 3 - 4 | 453 | 80 | |
| GenBank mRNA | AF100198.1 | - | 3 - 4 | 453 | 80 | |
| AK013945.1 | - | 3 - 4 | 336 | 80 | ||
| AK088026.1 | - | 3 - 4 | 336 | 80 | ||
| AK140930.1 | - | 3 - 4 | 334 | 80 | ||
| BC019945.1 | - | 3 - 4 | 322 | 3989811 | 80 | |
| BC052834.1 | - | 3 - 4 | 414 | 30048305 | 80 | |
| KF356400.1 | - | 2 - 3 | 238 | 80 | ||
| KF356401.1 | - | 2 - 3 | 238 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsdhl
Genomic Map