Assay Details
| Gene Symbol: Ntan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010946.3 | NP_035076.1 | 4 - 5 | 436 | 85 | |
| GenBank mRNA | AK041112.1 | - | 4 - 5 | 393 | 85 | |
| AK214245.1 | - | 2 - 3 | 184 | 85 | ||
| BC030172.1 | - | 4 - 5 | 434 | 5037501 | 85 | |
| U57692.1 | - | 4 - 5 | 390 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ntan1
Genomic Map