Assay Details
| Gene Symbol: Numb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136075.2 | NP_001129547.1 | 7 - 8 | 966 | 70 | |
| NM_001272055.1 | NP_001258984.1 | 6 - 7 | 933 | 70 | ||
| NM_001272056.1 | NP_001258985.1 | 8 - 9 | 1163 | 70 | ||
| NM_010949.2 | NP_035079.1 | 7 - 8 | 966 | 70 | ||
| NR_073563.1 | 9 - 10 | 1280 | 70 | |||
| GenBank mRNA | AF169191.1 | - | 6 - 7 | 918 | 70 | |
| AF169192.1 | - | 7 - 8 | 951 | 70 | ||
| AF170709.1 | - | 7 - 8 | 951 | 70 | ||
| AK004553.1 | - | 9 - 10 | 1188 | 70 | ||
| U70674.1 | - | 8 - 9 | 1084 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Numb
Genomic Map