Assay Details
| Gene Symbol: Numbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010950.2 | NP_035080.2 | 4 - 5 | 501 | 60 | |
| GenBank mRNA | AK147523.1 | - | 2 - 3 | 6288 | 60 | |
| BC059915.1 | - | 3 - 4 | 1090 | 60 | ||
| BC068116.1 | - | 4 - 5 | 328 | 6809443 | 60 | |
| BC098097.1 | - | 4 - 5 | 342 | 6403012 | 60 | |
| U96441.1 | - | 4 - 5 | 385 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Numbl
Genomic Map