Assay Details
| Gene Symbol: P4ha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136076.2 | NP_001129548.1 | 7 - 8 | 982 | 72 | |
| NM_011031.2 | NP_035161.2 | 7 - 8 | 982 | 72 | ||
| GenBank mRNA | AK053504.1 | - | 7 - 8 | 943 | 72 | |
| AK145535.1 | - | 7 - 8 | 909 | 72 | ||
| BC018411.1 | - | 7 - 8 | 894 | 3157481 | 72 | |
| U16163.1 | - | 6 - 7 | 869 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4ha2
Genomic Map