Assay Details
| Gene Symbol: Pkdrej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011105.2 | NP_035235.2 | 1 | 4152 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkdrej| Gene Symbol: Pkdrej | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011105.2 | NP_035235.2 | 1 | 4152 | 101 | |
Set Membership: |
|