Assay Details
| Gene Symbol: Pld3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011116.1 | NP_035246.1 | 12 - 13 | 1592 | 67 | |
| GenBank mRNA | AF026124.1 | - | 12 - 13 | 1592 | 67 | |
| AK153508.1 | - | 10 - 11 | 1281 | 67 | ||
| AK188372.1 | - | 1 - 2 | 150 | 67 | ||
| BC068170.1 | - | 12 - 13 | 1588 | 67 | ||
| BC076586.1 | - | 12 - 13 | 1656 | 30604186 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pld3
Genomic Map