Assay Details
| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011150.2 | NP_035280.1 | 1 - 2 | 142 | 75 | |
| GenBank mRNA | AK079915.1 | - | 1 - 2 | 171 | 75 | |
| AK137836.1 | - | 1 - 2 | 119 | 75 | ||
| AK150021.1 | - | 1 - 2 | 171 | 75 | ||
| AK152182.1 | - | 1 - 2 | 84 | 75 | ||
| AK153313.1 | - | 1 - 2 | 158 | 75 | ||
| AK153396.1 | - | 1 - 2 | 160 | 75 | ||
| AK155031.1 | - | 1 - 2 | 84 | 75 | ||
| BC090658.1 | - | 1 - 2 | 138 | 30666981 | 75 | |
| L16894.1 | - | 1 - 2 | 149 | 75 | ||
| X67809.1 | - | 1 - 2 | 142 | 75 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK079915.1,AK137836.1,AK150021.1,AK152182.1,AK153313.1,AK153396.1,AK155031.1,BC090658.1,L16894.1,NM_011150.2,X67809.1 transcripts.
Set Membership: |
|

Lgals3bp
Genomic Map