Assay Details
| Gene Symbol: Prl4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011165.3 | NP_035295.1 | 2 - 3 | 256 | 69 | |
| GenBank mRNA | AF011383.1 | - | 2 - 3 | 257 | 69 | |
| AF015562.1 | - | 2 - 3 | 256 | 69 | ||
| AK141285.1 | - | 2 - 3 | 268 | 69 | ||
| AK164009.1 | - | 2 - 3 | 268 | 69 | ||
| BC052218.1 | - | 2 - 3 | 257 | 30017285 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prl4a1
Genomic Map