Assay Details
| Gene Symbol: Ptgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011198.3 | NP_035328.2 | 3 - 4 | 414 | 96 | |
| GenBank mRNA | AK049923.1 | - | 3 - 4 | 405 | 96 | |
| AK144726.1 | - | 3 - 4 | 439 | 96 | ||
| AK144956.1 | - | 3 - 4 | 437 | 96 | ||
| AK149820.1 | - | 3 - 4 | 439 | 96 | ||
| AK166221.1 | - | 3 - 4 | 437 | 96 | ||
| AK172161.1 | - | 3 - 4 | 439 | 96 | ||
| M64291.1 | - | 3 - 4 | 438 | 96 | ||
| M88242.1 | - | 3 - 4 | 414 | 96 | ||
| M94967.1 | - | 3 - 4 | 439 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptgs2
Genomic Map