Assay Details
| Gene Symbol: Nek4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF099067.1 | - | 1 - 2 | 232 | 69 | |
| AJ223071.1 | - | 1 - 2 | 322 | 69 | ||
| AK078809.1 | - | 1 - 2 | 344 | 69 | ||
| AK132803.1 | - | 1 - 2 | 260 | 69 | ||
| AK144130.1 | - | 1 - 2 | 206 | 69 | ||
| AK147381.1 | - | 1 - 2 | 310 | 69 | ||
| AK149584.1 | - | 1 - 2 | 260 | 69 | ||
| AK168094.1 | - | 1 - 2 | 243 | 69 | ||
| BC057939.1 | - | 1 - 2 | 234 | 5325558 | 69 | |
| Y09234.1 | - | 1 - 2 | 322 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nek4
Genomic Map