Assay Details
| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286743.1 | NP_001273672.1 | 2 - 3 | 374 | 59 | |
| NM_001286744.1 | NP_001273673.1 | 2 - 3 | 280 | 59 | ||
| NM_011861.3 | NP_035991.1 | 2 - 3 | 339 | 59 | ||
| NM_178365.4 | NP_848142.1 | 2 - 3 | 357 | 59 | ||
| GenBank mRNA | AK043967.1 | - | 2 - 3 | 343 | 59 | |
| AK158686.1 | - | 2 - 3 | 377 | 59 | ||
| BC014698.1 | - | 2 - 3 | 228 | 4527708 | 59 | |
| CX200478.1 | - | 2 - 3 | 392 | 59 | ||
| X85124.1 | - | 2 - 3 | 248 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pacsin1
Genomic Map