Assay Details
| Gene Symbol: Pcbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011865.3 | NP_035995.1 | 1 | 585 | 100 | |
| GenBank mRNA | AF139894.1 | - | 1 | 354 | 100 | |
| AK002433.1 | - | 1 | 344 | 100 | ||
| BC004793.1 | - | 1 | 446 | 3588737 | 100 | |
| BC069915.1 | - | 1 | 585 | 3673570 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610523 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610525 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610519 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610527 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610513 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610524 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610528 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980326 | BC004793.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980326 | BC069915.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1980326 | NM_011865.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980326 | BC004793.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980326 | BC069915.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1980326 | NM_011865.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P241129 | BC004793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P241129 | BC069915.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P241129 | NM_011865.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P241129 | AF139894.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P241129 | AK002433.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P248544 | BC004793.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P248544 | BC069915.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P248544 | NM_011865.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P248544 | AF139894.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610506 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610499 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610524 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610544 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610537 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610529 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610532 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610547 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610542 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610498 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610535 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24610503 | - | - |
| Whole Mouse Genome G4122A | A_51_P248544 | BC004793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248544 | BC069915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248544 | NM_011865.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P248544 | AF139894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241129 | BC004793.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241129 | BC069915.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241129 | NM_011865.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241129 | AF139894.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P241129 | AK002433.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121531 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878980 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05591087 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05591072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05878976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610523 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610514 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610512 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610528 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610515 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610525 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610516 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610519 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610527 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610520 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610518 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610524 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610517 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24610513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610555 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24610466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05588693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05591060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05666201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05851397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05666200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05390781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610499 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610522 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05228858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610481 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610465 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05591063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610474 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05224272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05138810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05840366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05666194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05591080 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610484 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05591071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05352139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24610493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05477850 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10545920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10545919 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448642_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10545917 | - | - |
| Mouse Genome 430 2.0 Array | 1448642_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448642_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 23983_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 23983_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600020501.mm.1 | TC0600002639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020501.mm.1 | NM_011865.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020501.mm.1 | NM_011865.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020500.mm.1 | TC0600002639.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020500.mm.1 | NM_011865.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020500.mm.1 | NM_011865.3 | Refseq |
| Mouse Expression Set 430 | 1448642_PM_at | - | - |
| Mouse Expression Set 430 | 1448642_at | - | - |
| Murine Genome U74v2 Set | 94552_at | - | - |
| Murine Genome U74v2 Set | 94552_PM_at | - | - |
| Murine Genome U74v2 Set | 170769_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170769_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448642_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448642_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448642_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448642_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 784582 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1050088 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1050088 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001050088 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2702197 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2702197 | - | - |
| mouse-6 v1.1 (Illumina) | scl28749.2.254_68-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1050088 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28749.2.254_68-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1050088 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2702197 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2702197 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl28749.2.254_68-S | - | - |

Pcbp1
Genomic Map