Assay Details
| Gene Symbol: Plod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142916.1 | NP_001136388.1 | 12 - 13 | 1590 | 108 | |
| NM_011961.3 | NP_036091.2 | 12 - 13 | 1590 | 108 | ||
| GenBank mRNA | AF080572.1 | - | 12 - 13 | 1361 | 108 | |
| AK033363.1 | - | 12 - 13 | 1549 | 108 | ||
| AK046364.1 | - | 12 - 13 | 1594 | 108 | ||
| AK157207.1 | - | 12 - 13 | 1550 | 108 | ||
| BC021352.1 | - | 12 - 13 | 1422 | 5007509 | 108 | |
More Information
Set Membership: |
|

Plod2
Genomic Map