Assay Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011962.3 | NP_036092.1 | 9 - 10 | 1681 | 71 | |
| GenBank mRNA | AF046783.1 | - | 9 - 10 | 1089 | 71 | |
| AK013195.1 | - | 9 - 10 | 1227 | 71 | ||
| AK033360.1 | - | 9 - 10 | 1226 | 71 | ||
| AK053320.1 | - | 9 - 10 | 1323 | 71 | ||
| AK088948.1 | - | 9 - 10 | 1098 | 71 | ||
| BC043047.1 | - | 9 - 10 | 1680 | 5703967 | 71 | |
| BC054734.1 | - | 9 - 10 | 1681 | 5720686 | 71 | |
More Information
Set Membership: |
|

Plod3
Genomic Map