Assay Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011962.3 | NP_036092.1 | 12 - 13 | 2036 | 85 | |
| GenBank mRNA | AF046783.1 | - | 12 - 13 | 1444 | 85 | |
| AK013195.1 | - | 12 - 13 | 1582 | 85 | ||
| AK033360.1 | - | 12 - 13 | 1581 | 85 | ||
| AK053320.1 | - | 12 - 13 | 1678 | 85 | ||
| AK088948.1 | - | 12 - 13 | 1453 | 85 | ||
| BC043047.1 | - | 12 - 13 | 2035 | 5703967 | 85 | |
| BC054734.1 | - | 12 - 13 | 2036 | 5720686 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod3
Genomic Map