Assay Details
| Gene Symbol: Plod3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011962.3 | NP_036092.1 | 15 - 16 | 2362 | 66 | |
| GenBank mRNA | AF046783.1 | - | 15 - 16 | 1770 | 66 | |
| AK013195.1 | - | 15 - 16 | 1908 | 66 | ||
| AK033360.1 | - | 15 - 16 | 1907 | 66 | ||
| AK088948.1 | - | 15 - 16 | 1779 | 66 | ||
| BC043047.1 | - | 15 - 16 | 2361 | 5703967 | 66 | |
| BC054734.1 | - | 15 - 16 | 2362 | 5720686 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plod3
Genomic Map