Assay Details
| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012045.4 | NP_036175.2 | 2 - 3 | 293 | 127 | |
| GenBank mRNA | AF166099.2 | - | 2 - 3 | 292 | 127 | |
| AK029254.1 | - | 2 - 3 | 283 | 127 | ||
| BC125567.1 | - | 2 - 3 | 194 | 40130082 | 127 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF166099.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020952 | AK029254.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020952 | NM_012045.4 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 66923 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 551504 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103410 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165650 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179293 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020952 | AK029254.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020952 | NM_012045.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P114691 | AK029254.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P114691 | NM_012045.4 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347796 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347767 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347757 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347780 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347787 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347791 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347754 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347766 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347798 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347785 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347794 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347788 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347783 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347784 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347789 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347762 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347773 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347778 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347769 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347764 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347774 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347790 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347775 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347760 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23347756 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01002394 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179293 | - | - |
| Whole Mouse Genome G4122A | A_52_P114691 | AK029254.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P114691 | NM_012045.4 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107101 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179293 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03436938 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03436936 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03436935 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03436934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23347791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03413677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03376261 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04039805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04018529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347773 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03342129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347724 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03620435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03770520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347767 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347787 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03770509 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347777 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347737 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03304593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03784972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03998323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03342106 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347701 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03596211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04039802 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347770 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347794 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03945823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03342130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03342123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04094606 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347749 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03342121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347791 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23347729 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10517653 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517651 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517647 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517650 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517649 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517648 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517654 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10517652 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434852_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421325_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10517646 | - | - |
| Mouse Genome 430 2.0 Array | 1434852_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434852_at | - | - |
| Mouse Genome 430 2.0 Array | 1421325_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421325_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26971_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 26971_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400031576.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031576.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031576.mm.1 | NM_012045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031573.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031573.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031573.mm.1 | NM_012045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031572.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031572.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031572.mm.1 | NM_012045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031574.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031574.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031574.mm.1 | NM_012045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031577.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031577.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031577.mm.1 | NM_012045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031575.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031575.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031575.mm.1 | NM_012045.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031571.mm.1 | TC0400003829.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400031571.mm.1 | NM_012045.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400031571.mm.1 | NM_012045.4 | Refseq |
| Mouse Expression Set 430 | 1434852_at | - | - |
| Mouse Expression Set 430 | 1421325_at | - | - |
| Mouse Expression Set 430 | 1421325_PM_at | - | - |
| Mouse Expression Set 430 | 1434852_PM_at | - | - |
| Murine Genome U74v2 Set | 165529_r_at | - | - |
| Murine Genome U74v2 Set | 165529_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434852_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434852_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421325_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421325_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 405315 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120601 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120601 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120601 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230424 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230424 | - | - |
| mouse-6 v1.1 (Illumina) | scl23630.5_89-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120601 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23630.5_89-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120601 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1230424 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1230424 | - | - |

Pla2g2f
Genomic Map