Assay Details
| Gene Symbol: Ahr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013464.4 | NP_038492.1 | 5 - 6 | 926 | 90 | |
| GenBank mRNA | AF405560.1 | - | 5 - 6 | 560 | 90 | |
| AF405561.1 | - | 5 - 6 | 560 | 90 | ||
| AF405562.1 | - | 5 - 6 | 560 | 90 | ||
| AF405563.1 | - | 5 - 6 | 560 | 90 | ||
| AF405565.1 | - | 5 - 6 | 560 | 90 | ||
| AF405566.1 | - | 5 - 6 | 560 | 90 | ||
| AF405567.1 | - | 5 - 6 | 560 | 90 | ||
| AF405568.1 | - | 5 - 6 | 560 | 90 | ||
| AF405570.1 | - | 5 - 6 | 560 | 90 | ||
| AK153678.1 | - | 5 - 6 | 927 | 90 | ||
| AK155380.1 | - | 5 - 6 | 978 | 90 | ||
| AY662080.1 | - | 5 - 6 | 560 | 90 | ||
| AY662081.1 | - | 4 - 5 | 456 | 90 | ||
| AY662082.1 | - | 5 - 6 | 560 | 90 | ||
| AY662083.1 | - | 5 - 6 | 560 | 90 | ||
| AY662084.1 | - | 5 - 6 | 560 | 90 | ||
| D38416.1 | - | 5 - 6 | 575 | 90 | ||
More Information
Set Membership: |
|

Ahr
Genomic Map