Assay Details
| Gene Symbol: Ahr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013464.4 | NP_038492.1 | 8 - 9 | 1365 | 74 | |
| GenBank mRNA | AF405563.1 | - | 8 - 9 | 999 | 74 | |
| AK136072.1 | - | 2 - 3 | 249 | 74 | ||
| AK153678.1 | - | 8 - 9 | 1366 | 74 | ||
More Information
Set Membership: |
|

Ahr
Genomic Map