Assay Details
| Gene Symbol: Anxa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013469.2 | NP_038497.2 | 4 - 5 | 702 | 78 | |
| GenBank mRNA | AK044203.1 | - | 4 - 5 | 675 | 78 | |
| AK137158.1 | - | 4 - 5 | 646 | 78 | ||
| AK140375.1 | - | 4 - 5 | 675 | 78 | ||
| BC012875.1 | - | 4 - 5 | 622 | 3500318 | 78 | |
| CT010331.1 | - | 3 - 4 | 553 | 78 | ||
| U65986.1 | - | 4 - 5 | 687 | 78 | ||
More Information
Set Membership: |
|

Anxa11
Genomic Map