Assay Details
| Gene Symbol: Phtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163467.1 | NP_001156939.1 | 11 - 12 | 1427 | 70 | |
| NM_001163468.1 | NP_001156940.1 | 10 - 11 | 1674 | 70 | ||
| NM_001163469.1 | NP_001156941.1 | 10 - 11 | 1698 | 70 | ||
| NM_013629.2 | NP_038657.2 | 11 - 12 | 1833 | 70 | ||
| GenBank mRNA | AJ133721.1 | - | 11 - 12 | 1427 | 70 | |
| AJ242864.1 | - | 11 - 12 | 1833 | 70 | ||
| AK019146.1 | - | 11 - 12 | 1737 | 70 | ||
| AK088714.1 | - | 10 - 11 | 1565 | 70 | ||
| AK131629.1 | - | 1724 | 70 | |||
| BC021147.1 | - | 11 - 12 | 1706 | 70 | ||
| BC050825.1 | - | 10 - 11 | 1510 | 6518647 | 70 | |
| BC057340.1 | - | 11 - 12 | 1719 | 6827816 | 70 | |
| BC060177.1 | - | 11 - 12 | 1719 | 6827816 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phtf1
Genomic Map