Assay Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163565.1 | NP_001157037.1 | 5 - 6 | 787 | 109 | |
| NM_013643.2 | NP_038671.2 | 5 - 6 | 895 | 109 | ||
| GenBank mRNA | AK038416.1 | - | 5 - 6 | 564 | 109 | |
| BC051975.1 | - | 5 - 6 | 536 | 109 | ||
| BC057339.1 | - | 4 - 5 | 1057 | 109 | ||
| BC079592.1 | - | 5 - 6 | 867 | 6817360 | 109 | |
| S80329.1 | - | 2 - 3 | 731 | 109 | ||
| U28216.1 | - | 5 - 6 | 648 | 109 | ||
| U28217.1 | - | 5 - 6 | 906 | 109 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the S80329.1 transcripts.
Set Membership: |
|

Ptpn5
Genomic Map