Assay Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163565.1 | NP_001157037.1 | 9 - 10 | 1384 | 82 | |
| NM_013643.2 | NP_038671.2 | 9 - 10 | 1492 | 82 | ||
| GenBank mRNA | AK038416.1 | - | 9 - 10 | 1161 | 82 | |
| BC051975.1 | - | 9 - 10 | 1133 | 82 | ||
| BC057339.1 | - | 8 - 9 | 1654 | 82 | ||
| BC079592.1 | - | 9 - 10 | 1464 | 6817360 | 82 | |
| U28217.1 | - | 9 - 10 | 1503 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn5
Genomic Map