Assay Details
| Gene Symbol: Ptpn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163565.1 | NP_001157037.1 | 11 - 12 | 1632 | 73 | |
| NM_013643.2 | NP_038671.2 | 11 - 12 | 1740 | 73 | ||
| GenBank mRNA | AK038416.1 | - | 11 - 12 | 1409 | 73 | |
| BC051975.1 | - | 11 - 12 | 1381 | 73 | ||
| BC057339.1 | - | 10 - 11 | 1902 | 73 | ||
| U28217.1 | - | 11 - 12 | 1751 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn5
Genomic Map