Assay Details
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048146.2 | NP_001041611.1 | 5 - 6 | 1034 | 63 | |
| NM_001286507.1 | NP_001273436.1 | 4 - 5 | 934 | 63 | ||
| NM_001286508.1 | NP_001273437.1 | 5 - 6 | 952 | 63 | ||
| NM_013727.4 | NP_038755.1 | 5 - 6 | 1034 | 63 | ||
| GenBank mRNA | AB007141.1 | - | 5 - 6 | 870 | 63 | |
| AK004992.1 | - | 5 - 6 | 1013 | 63 | ||
| AK015942.1 | - | 5 - 6 | 855 | 63 | ||
| AK030076.1 | - | 5 - 6 | 866 | 63 | ||
| AK036102.1 | - | 4 - 5 | 841 | 63 | ||
| AK134714.1 | - | 5 - 6 | 937 | 63 | ||
| AK153477.1 | - | 5 - 6 | 998 | 63 | ||
| AK165549.1 | - | 5 - 6 | 920 | 63 | ||
| AK167783.1 | - | 5 - 6 | 978 | 63 | ||
| BC006851.1 | - | 5 - 6 | 929 | 3156857 | 63 | |
More Information
Set Membership: |
|

Azi2
Genomic Map