Assay Details
| Gene Symbol: Nufip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013745.5 | NP_038773.1 | 6 - 7 | 842 | 77 | |
| GenBank mRNA | AF159549.1 | - | 6 - 7 | 800 | 77 | |
| AK032324.1 | - | 6 - 7 | 843 | 77 | ||
| AK156205.1 | - | 6 - 7 | 829 | 77 | ||
| BC020364.1 | - | 6 - 7 | 817 | 77 | ||
| BC056192.1 | - | 6 - 7 | 828 | 30024494 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nufip1
Genomic Map