Assay Details
| Gene Symbol: Adamts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013906.2 | NP_038934.2 | 7 - 8 | 2243 | 66 | |
| GenBank mRNA | AF175282.1 | - | 7 - 8 | 2246 | 66 | |
| AK043875.1 | - | 1215 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts8
Genomic Map