| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100023736.mm.1 |
NM_001131054.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023736.mm.1 |
NM_001131054.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023736.mm.1 |
NM_013917.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023736.mm.1 |
NM_013917.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023736.mm.1 |
TC1100002567.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023738.mm.1 |
NM_013917.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023738.mm.1 |
NM_013917.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023738.mm.1 |
TC1100002567.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023734.mm.1 |
NM_001131054.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023734.mm.1 |
NM_001131054.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023734.mm.1 |
NM_013917.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023734.mm.1 |
NM_013917.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023734.mm.1 |
TC1100002567.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023739.mm.1 |
NM_013917.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023739.mm.1 |
NM_013917.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023739.mm.1 |
TC1100002567.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023732.mm.1 |
NM_001131054.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023732.mm.1 |
NM_001131054.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023732.mm.1 |
NM_013917.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023732.mm.1 |
NM_013917.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023732.mm.1 |
TC1100002567.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100023735.mm.1 |
NM_001131054.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023735.mm.1 |
NM_001131054.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023735.mm.1 |
NM_013917.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023735.mm.1 |
NM_013917.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100023735.mm.1 |
TC1100002567.mm.1 |
MTA TC |