Assay Details
| Gene Symbol: Hacd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013935.3 | NP_038963.3 | 4 - 5 | 530 | 85 | |
| GenBank mRNA | AK010119.1 | - | 4 - 5 | 375 | 85 | |
| BC138116.1 | - | 5 - 6 | 518 | 8861135 | 85 | |
| BC141017.1 | - | 5 - 6 | 518 | 9055629 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hacd1
Genomic Map