Assay Details
| Gene Symbol: Hspa14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015765.2 | NP_056580.2 | 9 - 10 | 1009 | 88 | |
| GenBank mRNA | AK031489.1 | - | 948 | 88 | ||
| AK145937.1 | - | 9 - 10 | 963 | 88 | ||
| AK153397.1 | - | 9 - 10 | 971 | 88 | ||
| BC002056.1 | - | 9 - 10 | 937 | 3493119 | 88 | |
| BI734146.1 | - | 1 - 2 | 112 | 88 | ||
| BU505607.1 | - | 6 - 7 | 660 | 88 | ||
| U08215.1 | - | 9 - 10 | 969 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hspa14
Genomic Map