Assay Details
| Gene Symbol: Ppap2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005452.1 | - | 1 - 2 | 121 | 81 | |
| AK049581.1 | - | 1 - 2 | 164 | 81 | ||
| BC010332.1 | - | 1 - 2 | 142 | 2654393 | 81 | |
| BY344892.1 | - | 1 - 2 | 167 | 81 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AF123611.1 | - | 156 | Chr. - - [-] on Build | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277402 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277401 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277400 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129437 | BB737982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2129437 | NM_015817.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631736 | AK052443.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631736 | DQ286956.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631736 | NM_001103168.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631736 | XM_001475156.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631736 | XM_001475715.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 860003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377130 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680868 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114609 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129437 | BB737982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2129437 | NM_015817.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P607674 | BC085104.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P607674 | AK208995.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P607674 | BC010332.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P607674 | BC016635.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P607674 | NM_015817.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277437 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277439 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277436 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277403 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277427 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277399 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277418 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277419 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277417 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277430 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277434 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277393 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27277407 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P114609 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07119744 | - | - |
| Whole Mouse Genome G4122A | A_52_P607674 | BC085104.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P607674 | AK208995.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P607674 | BC010332.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P607674 | BC016635.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P607674 | NM_015817.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107801 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134696 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07119744 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P114609 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08838777 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08838799 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08838774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08838773 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08838793 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08838794 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277400 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277399 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277402 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277398 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27277401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277342 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277393 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27277358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277424 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08748493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277437 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277406 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08430398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277412 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277414 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08733139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08543820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08898055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08898047 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08277860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08430410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08223190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277427 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08838799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08543825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277419 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08223202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27277421 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08277859 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10370553 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370556 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370555 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370557 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370554 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10370558 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451210_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420910_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10370552 | - | - |
| Mouse Genome 430 2.0 Array | 1420910_at | - | - |
| Mouse Genome 430 2.0 Array | 1451210_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451210_at | - | - |
| Mouse Genome 430 2.0 Array | 1420910_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50784_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 50784_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000017968.mm.1 | NM_015817.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017968.mm.1 | TC1000002467.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017969.mm.1 | NM_015817.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017969.mm.1 | TC1000002467.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017967.mm.1 | NM_015817.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017967.mm.1 | TC1000002467.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000017970.mm.1 | NM_015817.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000017970.mm.1 | TC1000002467.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420910_PM_at | - | - |
| Mouse Expression Set 430 | 1420910_at | - | - |
| Mouse Expression Set 430 | 1451210_PM_at | - | - |
| Mouse Expression Set 430 | 1451210_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420910_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451210_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451210_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420910_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420910_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420910_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451210_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451210_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 107100711 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2970348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107100711 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002970348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2655746 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2655746 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1249666 | - | - |
| mouse-6 v1.1 (Illumina) | scl37761.6_244-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0050784.2_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2970348 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107100711 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0050784.2_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107100711 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3003631 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249666 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1216282 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3003631 | - | - |

Ppap2c
Genomic Map