Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 2 - 3 | 284 | 75 | |
| AK018166.1 | - | 3 - 4 | 335 | 75 | ||
| AK086301.1 | - | 3 - 4 | 494 | 75 | ||
| AK134581.1 | - | 3 - 4 | 338 | 75 | ||
| AK153400.1 | - | 3 - 4 | 322 | 75 | ||
| BC005640.1 | - | 3 - 4 | 323 | 75 | ||
| BC018178.1 | - | 3 - 4 | 309 | 75 | ||
| BC037451.1 | - | 2 - 3 | 313 | 4018648 | 75 | |
| U27462.1 | - | 3 - 4 | 531 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nub1
Genomic Map