Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 10 - 11 | 1242 | 62 | |
| AK018166.1 | - | 10 - 11 | 2051 | 62 | ||
| AK086301.1 | - | 11 - 12 | 1452 | 62 | ||
| AK134581.1 | - | 11 - 12 | 1296 | 62 | ||
| U27462.1 | - | 11 - 12 | 1489 | 62 | ||
More Information
Set Membership: |
|

Nub1
Genomic Map