Assay Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF534114.1 | - | 12 - 13 | 1485 | 104 | |
| AK018166.1 | - | 12 - 13 | 2294 | 104 | ||
| AK086301.1 | - | 13 - 14 | 1695 | 104 | ||
| AK134581.1 | - | 13 - 14 | 1539 | 104 | ||
| AK153400.1 | - | 13 - 14 | 1519 | 104 | ||
| BC037451.1 | - | 12 - 13 | 1514 | 4018648 | 104 | |
| U27462.1 | - | 13 - 14 | 1732 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nub1
Genomic Map