Assay Details
| Gene Symbol: Nell2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289653.1 | NP_001276582.1 | 3 - 4 | 234 | 66 | |
| GenBank mRNA | AK035019.1 | - | 1 - 2 | 87 | 66 | |
| AK039790.1 | - | 298 | 66 | |||
| AK048913.1 | - | 505 | 66 | |||
| BC051968.1 | - | 2 - 3 | 202 | 6401465 | 66 | |
| BU612839.1 | - | 3 - 4 | 443 | 66 | ||
| U59230.1 | - | 3 - 4 | 235 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nell2
Genomic Map