Assay Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016784.3 | NP_058064.2 | 3 - 4 | 365 | 76 | |
| GenBank mRNA | AF044334.1 | - | 2 - 3 | 257 | 76 | |
| AK075993.1 | - | 3 - 4 | 353 | 76 | ||
| AK076007.1 | - | 3 - 4 | 343 | 76 | ||
| AK077197.1 | - | 3 - 4 | 366 | 76 | ||
| AK080068.1 | - | 3 - 4 | 372 | 76 | ||
| BC006750.1 | - | 3 - 4 | 335 | 3709134 | 76 | |
| BC052895.1 | - | 3 - 4 | 323 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plrg1
Genomic Map