Assay Details
| Gene Symbol: Bysl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016859.3 | NP_058555.3 | 6 - 7 | 1290 | 77 | |
| GenBank mRNA | AF007802.1 | - | 4 - 5 | 519 | 77 | |
| AK019239.1 | - | 6 - 7 | 1025 | 77 | ||
| AK032993.1 | - | 6 - 7 | 1022 | 77 | ||
| AK049195.1 | - | 6 - 7 | 1283 | 77 | ||
| AK051704.1 | - | 6 - 7 | 1280 | 77 | ||
| AK083033.1 | - | 6 - 7 | 1277 | 77 | ||
| AK143027.1 | - | 6 - 7 | 1291 | 77 | ||
| AK168301.1 | - | 6 - 7 | 1028 | 77 | ||
| BC017530.1 | - | 6 - 7 | 1017 | 4925307 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213166 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 363901 | AK082252.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621636 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29797 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100538 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 221905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 67123 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135231 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153281 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P269396 | AK083033.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P269396 | AK143027.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P269396 | NM_016859.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P447946 | AK168301.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P447946 | BC017530.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P447946 | AK083033.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P447946 | AK143027.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P447946 | AK049195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213137 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213157 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135231 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153281 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08792835 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08792850 | - | - |
| Whole Mouse Genome G4122A | A_52_P269396 | AK083033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P269396 | AK143027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P269396 | NM_016859.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK168301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | BC017530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK083033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK049195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P131836 | AK051704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447946 | AK168301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447946 | BC017530.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447946 | AK083033.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447946 | AK143027.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P447946 | AK049195.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140080 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153281 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135231 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583798 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583795 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583800 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13583789 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213027 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213132 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213124 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13461766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213142 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13436543 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213122 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213125 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13156650 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13498316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213150 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213129 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13278572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213130 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213136 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213144 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213162 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213094 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213097 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213164 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213146 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13316448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213165 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13564307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213121 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13278579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213134 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213155 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451585 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451583 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451587 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451590 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451588 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450742_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422767_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451580 | - | - |
| Mouse Genome 430 2.0 Array | 1450742_at | - | - |
| Mouse Genome 430 2.0 Array | 1450742_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422767_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422767_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53414_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53414_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700020849.mm.1 | NM_016859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020849.mm.1 | NM_016859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020849.mm.1 | TC1700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020851.mm.1 | NM_016859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020851.mm.1 | NM_016859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020851.mm.1 | TC1700002208.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700020850.mm.1 | NM_016859.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020850.mm.1 | NM_016859.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700020850.mm.1 | TC1700002208.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1422767_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450742_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450742_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422767_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3170110 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170110 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2698719 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2698719 | - | - |
| mouse-6 v1.1 (Illumina) | scl49839.8_365-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170110 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49839.8_365-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170110 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2698719 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2698719 | - | - |

Bysl
Genomic Map