Assay Details
| Gene Symbol: Pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017463.2 | NP_059491.1 | 2 - 3 | 373 | 65 | |
| GenBank mRNA | AF020198.1 | - | 2 - 3 | 375 | 65 | |
| AK134357.1 | - | 2 - 3 | 294 | 65 | ||
| AK141763.1 | - | 297 | 65 | |||
| AK159623.1 | - | 2 - 3 | 582 | 65 | ||
| BC010287.1 | - | 3 - 4 | 485 | 2599030 | 65 | |
| BC062147.1 | - | 2 - 3 | 549 | 6505014 | 65 | |
| BC087615.1 | - | 2 - 3 | 353 | 4978305 | 65 | |
| CT010391.1 | - | 2 - 3 | 290 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pbx2
Genomic Map