Assay Details
| Gene Symbol: Pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017463.2 | NP_059491.1 | 7 - 8 | 1203 | 54 | |
| GenBank mRNA | AF020198.1 | - | 7 - 8 | 1205 | 54 | |
| AK134357.1 | - | 7 - 8 | 1124 | 54 | ||
| AK141763.1 | - | 1127 | 54 | |||
| AK159623.1 | - | 7 - 8 | 1412 | 54 | ||
| BC010287.1 | - | 8 - 9 | 1315 | 2599030 | 54 | |
| BC062147.1 | - | 7 - 8 | 1379 | 6505014 | 54 | |
| BC087615.1 | - | 7 - 8 | 1183 | 4978305 | 54 | |
| CT010391.1 | - | 7 - 8 | 1120 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pbx2
Genomic Map