Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 4 - 5 | 432 | 64 | |
| GenBank mRNA | AB037111.1 | - | 5 - 6 | 717 | 64 | |
| AB037181.1 | - | 4 - 5 | 432 | 64 | ||
| AK034493.1 | - | 4 - 5 | 664 | 64 | ||
| AK041937.1 | - | 4 - 5 | 621 | 64 | ||
| AK046540.1 | - | 4 - 5 | 626 | 64 | ||
| AK047692.1 | - | 4 - 5 | 620 | 64 | ||
| AK080951.1 | - | 4 - 5 | 609 | 64 | ||
| AK166100.1 | - | 4 - 5 | 680 | 64 | ||
| BC137898.1 | - | 4 - 5 | 415 | 8860917 | 64 | |
| BC145016.1 | - | 4 - 5 | 418 | 9053564 | 64 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB037111.1,AB037181.1,AK047692.1,AK080951.1,AK166100.1,BC137898.1,BC145016.1,NM_018830.1 transcripts.
Set Membership: |
|

Asah2
Genomic Map