Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 12 - 13 | 1508 | 89 | |
| GenBank mRNA | AB037111.1 | - | 13 - 14 | 1793 | 89 | |
| AB037181.1 | - | 12 - 13 | 1508 | 89 | ||
| AK034493.1 | - | 12 - 13 | 1740 | 89 | ||
| AK046540.1 | - | 12 - 13 | 1702 | 89 | ||
| AK080951.1 | - | 12 - 13 | 1685 | 89 | ||
| BC022604.1 | - | 3 - 4 | 290 | 89 | ||
| BC137898.1 | - | 12 - 13 | 1491 | 8860917 | 89 | |
| BC145016.1 | - | 12 - 13 | 1494 | 9053564 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asah2
Genomic Map