Assay Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018830.1 | NP_061300.1 | 18 - 19 | 2038 | 87 | |
| GenBank mRNA | AB037111.1 | - | 19 - 20 | 2323 | 87 | |
| AB037181.1 | - | 18 - 19 | 2038 | 87 | ||
| BC022604.1 | - | 9 - 10 | 820 | 87 | ||
| BC137898.1 | - | 18 - 19 | 2021 | 8860917 | 87 | |
| BC145016.1 | - | 18 - 19 | 2021 | 9053564 | 87 | |
More Information
Set Membership: |
|

Asah2
Genomic Map